GMF syringe filters, Target®
Leverandør: THERMO NATIONAL SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F2500-18 F2500-19 F2500-20
NSCAF2500-18EA 3000 DKK
NSCAF2500-18 NSCAF2500-19 NSCAF2500-20
GMF syringe filters, Target®
Filtre Sprøjtefiltre
The Target® syringe filters are ideal for general purpose laboratory use. Target2® GMF (Glass MicroFiber) syringe filters are used for dissolution testing. They are also well-suited for general filtration applications.
  • Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample
  • Performance-tested under actual HPLC conditions
  • 30 mm products are pressure rated to 120 psi (8,3 bar)
  • Able to filter solutions at temperatures up to 100 °C
Housings are manufactured from solvent-resistant, low-extractable PP resins specifically selected for wide compatibility with common HPLC sample matrices.