Chemical resistant gloves, Butoject® 898
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
898.11 898.8 898.10 898.9
112-1367EA 452 DKK
112-1367 112-4368 112-4367 112-4369
Chemical resistant gloves, Butoject® 898
Handsker
These highly flexible, semi-disposable chemical protective gloves made of Butyl rubber are suitable for extreme demands when working with esters and ketones. Manufactured using the modern Vulcoject-process technology (injection moulding), this glove is particularly gas tight.
  • Flexible at low temperatures, good ozone resistance: Can be used under extreme climatic conditions
  • Very good protection against extremely aggressive chemicals
  • High tightness against gas (meets NATO requirements)
  • Powdered, smooth finish, rolled edge
  • Wide shape makes it possible to wear gloves underneath
Certificeringer: EN 388 (2111X), EN ISO 374-1 Type A (BCIKLM), EN 421; Cat. III; AQL < 0,65
Advarsel: May only be reused for certain chemical groups after a special cleaning process. Please contact your local Avantor customer service center for further information.