Chemical resistant gloves, Camatril® 730
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
730.8 730.9 730.10 730.11 730.7
KCHL730.8EA 692 DKK
KCHL730.8 KCHL730.9 KCHL730.10 KCHL730.11 KCHL730.7
Chemical resistant gloves, Camatril® 730
Handsker
These flock lined nitrile gloves provide high mechanical and chemical resistance. The high abrasion, cut and puncture resistance allows working with sharp edged materials.
  • Good grip when handling greasy and oily parts
  • Good temperature flexibility
  • Glove surface free from silicone (lacquer indifference, test method automotive industry)
  • Anti-static and electrostatic properties
  • Flock lining offers additional wearing comfort
Ideal for use in food processing, laboratories, in the automotive/petrochemistry/printing/chemical industry and in paint shops. Also suitable for the handling of epoxy resins.
Thickness: 0,4 ± 0,05 mm or 0,5 ± 0,1
Colour: Green
EN 388, EN 374 (AJL); Cat. III; AQL 0,65 Approved for food contact
Certificeringer: EN 374-1 Type A (AJKLOT), EN 388 (3001X); Cat. III; AQL <0,65
Food contact approved