Dialysis membranes, Spectra/Por® 6
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1891.1 1892.1 1893.1
ROTH1891.1EA 0 DKK
ROTH1891.1 ROTH1892.1 ROTH1893.1
Dialysis membranes, Spectra/Por® 6
Slanger Dialyseslanger
Dialysis tubing is made from cellulose membrane. It contains low amounts of impurities such as sulphides and heavy metals.
  • Stored at 4 °C
  • Temperature stable up to 60 °C
Emballering: Packaged wet in 0,05% sodium azide solution; on rolls.