Immersion coolers, with natural refrigerant, TC range
Leverandør: HUBER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3003.0002.99 3003.0004.99 3004.0002.99 3004.0004.99 3005.0046.99 3005.0043.99 3005.0044.99 3005.0045.99 3005.0107.99 3004.0020.00 3004.0019.00 3003.0044.00
HUBE3003.0002.99EA 28910 DKK
HUBE3003.0002.99 HUBE3003.0004.99 HUBE3004.0002.99 HUBE3004.0004.99 HUBE3005.0046.99 HUBE3005.0043.99 HUBE3005.0044.99 HUBE3005.0045.99 HUBE3005.0107.99 HUBE3004.0020.00 HUBE3004.0019.00 HUBE3003.0044.00
Immersion coolers, with natural refrigerant, TC range
Cirkulatorsystemer Lukkede cirkulatorsystemer Køletermostatbade
The TC range of immersion coolers are a flexible solution for a variety of cooling tasks. They are universally used for rapid cooling of fluids. Typical applications include counter-cooling for heat circulation thermostats or replacing cooling by dry ice, they are also an environmentally friendly alternative to mains water cooling. The TC immersion coolers are suitable for unsupervised use, for cooling tasks which require continuous cooling or as a variant with temperature control and a PT100 probe.
  • Provide rapid cooling of fluids
  • Ideal for counter-cooling or as a replacement for dry ice
  • LED temperature display (E models)
  • Easy operation by means of only three keys
  • Compact, space-saving construction
  • High quality stainless steel immersion probe
The models with suffix 'E' have an LED display with entry of nominal values and regulate with a precision of ±0,5 K. A spiral-shaped or flexible immersion cooling probe made out of stainless steel is available for all models so that fluids can be cooled in virtually all vessel types. The compact housing is made from high quality stainless steel, and is fitted with carrying grips. Units have a small footprint, require little installation area and optional accessory castors enable easy relocation around the laboratory.
The TC100 models use environmentally friendly refrigerant R1270 and other models use R290.
Bestillingsoplysninger: Accessories included. There are also some optional accessories.
Advarsel: Note: Cooling capacity at 0; –20; –30; –40; −50; −60; −80; –90; −100 °C.