Stand-up pouches with zip closure, Lamizip
Leverandør: DAKLAPACK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
338 356 358 354 353 352 621 357 337 355 332 350 301 336
300-0222EA 3100 DKK
300-0222 300-0324 300-0325 300-0323 300-0322 300-0321 216-1379 216-1378 216-1376 216-1377 216-1375 300-0320 DAKL301 DAKL336
Stand-up pouches with zip closure, Lamizip
Poser Prøveposer
These stand-up pouches are suitable for products which need special requirements for packaging such as food, drink or pharmaceutical products. They can be used for the packaging of samples, substances and preparations.
  • Laminated foil with high barrier properties against oxygen and water vapour
  • Can be opened and closed innumerable times
  • Offer protection against the effects of UV radiation
  • Round bottom for increased stability