Industrial balance, Defender™ 2000
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30235937 30235922 30235927 30235932 30235942 30235947 30235952 30235957
OHAU30235937EA 4330 DKK
OHAU30235937 OHAU30235922 OHAU30235927 OHAU30235932 OHAU30235942 OHAU30235947 OHAU30235952 OHAU30235957
Industrial balance, Defender™ 2000
Vægte
The Defender 2000 scale is designed for durability in commercial and industrial weighing applications. Software included for parts counting mode with selectable sample sizes and weighing totalisation (accumulation) functions. Selectable span calibration points, averaging level, auto-zero tracking and battery saving auto-off. Non-slip rubber levelling feet with locking nuts, externally visible level bubble.
  • Backlit display
  • Internal rechargeable battery
Economical scale features a sturdy, welded steel frame, polished stainless steel pan, and indicators with tactile keys.