Portable balances, gold pocket scale, YA series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80251911 80251910 80251912
611-5360EA 344 DKK
611-5360 611-5359 611-5361
Portable balances, gold pocket scale, YA series
Vægte
The unique YA series weighing pan is integrated right into its carrying case, offering the ultimate combination of portability and protection. Just flip the case open and begin weighing.
  • A hinged cover, sturdy plastic housing and stainless steel weighing pan protect the small balance from damage
  • With an extremely fast stabilisation time, your results will appear quickly on the blue backlit LCD display
  • Powered by two 'AAA' batteries, a power-saving auto shut-off feature helps to conserve battery life