Industrial scales, Explorer®
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30057068 30057069 30078073 30078074 30078075 30078076 30057067 30078072 30078077 30100518 30100519 30100520
611-4974EA 49620 DKK
611-4974 611-4975 611-4977 611-4978 OHAU30078075 OHAU30078076 611-4973 611-4976 OHAU30078077 OHAU30100518 OHAU30100519 OHAU30100520
Industrial scales, Explorer®
Vægte
Large platform bench scales that incorporate touch screen, advanced weighing and proximity sensor technologies. The easy to use control unit can be mounted on the base, above the base on an optional column, on a wall, or remotely (up to 3 metres distance with an optional cable). The touch screen display, with universal icons and informative prompts, simplifies set-up functions and basic operation. The operator has the added choice to use proximity sensors for hands-free operation for handling sensitive samples. The balances have multiple application modes for basic weighing to SQC applications together with short stabilisation times and optimised performance specifications.
  • Colour touch screen display with viewing angle adjustment in a demountable control unit
  • Advanced weighing technology with stabilisation times of approximately 1 second
  • Icon-driven menu for ease of navigation to required functionality
  • AutoCal™ internal calibration
  • Dual multi-functional proximity sensors for hands-free operation of basic or user selectable functions
  • LIMS compatibility
The weighing platform base (not control/display unit) has IP 54 protection rating.