Centrifuge tubes, round bottom, KIMBLE®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SP45500-15 SP45500-30 SP45600-15 SP45600-30
SCERSP45500-15EA 1350 DKK
SCERSP45500-15 SCERSP45500-30 SCERSP45600-15 SCERSP45600-30
Centrifuge tubes, round bottom, KIMBLE®
Rør Centrifugerør
Borosilicate glass, Type 1, Class B.
  • Manufactured from chemically strengthened borosilicate glass
  • Can be centrifuged up to 13100×g when used with the accessory rubber adapter sleeve in a 50 ml rotor cavity
  • Withstand temperatures up to 300 °C
  • Screw-thread tubes are supplied with PTFE-faced rubber liner caps
Bestillingsoplysninger: Accessories sleeves and replacement caps sold separately.