External national pipe thread adapters, Upchurch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
U-510 U-510-01 U-511 U-500 U-500-01 U-501 U-504
554-3266EA 307 DKK
554-3266 554-3267 UPCHU-511 554-3262 554-3263 UPCHU-501 UPCHU-504
External national pipe thread adapters, Upchurch Scientific®
Fittings og stik
These PEEK adapters make connections to female ¹/₈" and ¹/₄" National Pipe Thread (NPT) ports.
  • Chemically resistant
  • Pressure rating up to 500 psi (34 bar)