Metric threaded adapters, Upchurch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P-621 P-626 P-626-01 P-650 P-670 P-694
UPCHP-621EA 107 DKK
UPCHP-621 554-3126 554-3127 554-3136 UPCHP-670 554-3154
Metric threaded adapters, Upchurch Scientific®
Fittings og stik
These female M6 adapters are made from PEEK, EFTE or PCTFE.
  • Pressure rating up to 1000 psi (69 bar)