English threaded adapters, Upchurch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P-135 P-627 P-627-01 P-681 U-659
554-3093EA 292 DKK
554-3093 554-3128 554-3129 554-3149 UPCHU-659
English threaded adapters, Upchurch Scientific®
Fittings og stik
These adapters are made from PEEK or PCTFE.
  • Pressure rating up to 1000 psi (69 bar)