Chromatography tubing, FEP, Upchurch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1548L 1548XL 1520L 1520XL 1521L 1521XL 1524L 1524XL 1527L 1650L 1671L 1673L 1677L
554-2985EA 1880 DKK
554-2985 554-2987 554-2962 554-2963 554-2969 554-2974 UPCH1524L 554-2977 UPCH1527L UPCH1650L UPCH1671L UPCH1673L UPCH1677L
Chromatography tubing, FEP, Upchurch Scientific®
Slanger Kromatografirør
FEP tubing, coloured.
  • Features virtually identical chemical inertness when compared with PTFE
  • Some sizes available in multiple, translucent colours for easy flow path identification
  • Some sizes also in black for light-sensitive applications
This tubing is chemically inert and suitable for lower pressure applications.
1/4" OD tubing holds to 250 psi (17,2 bar); inner and outer diameter tolerances are ±0,004".
1/8" OD tubing holds to 500 psi (34 bar); inner and outer diameter tolerances are ±0,003".
1/16" OD tubing holds from 1000 psi (69 bar)More Product Information