Chemical resistant gloves, Ultranitril 480
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34480177 34480178 34480009 34480170
112-5589EA 96.9 DKK
112-5589 112-5590 112-5591 112-5592
Chemical resistant gloves, Ultranitril 480
Handsker
These nitrile gloves with chlorinated interior finish provide excellent protection during the intensive handling of high-risk chemical products.
  • Optimised service life: excellent mechanical resistance (abrasion, perforation)
  • Good grip on slippery objects, due to the embossed texture
  • Chlorinated finish makes the gloves easy to put on and pull off
  • Nitrile offers very good resistance to oils, greases and hydrocarbon derivatives
Ideal for treating/degreasing metals with aggressive solvents, handling drums of chemical products, timber treatment and finishing, application of pesticides and cleaning of printing-press rollers.
Thickness: 0,55 mm
Colour: Green
EN 3888 (4102), EN 374 (AJKL); Cat. III
Certificeringer: EN 388:2016 (4102X), EN ISO 374-1 Type A (AJKOPT), EN 374-5; ISO 18889 G2; Cat. III