Industrial gloves, Titan 375 / 376
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34375016 34375018 34375017 34375019
112-5566EA 693 DKK
112-5566 112-5568 112-5567 112-5569
Industrial gloves, Titan 375 / 376
Handsker
These nitrile gloves with textile lining and smooth finish provide protection against humidity, oil and grease splashes.
  • Good abrasion resistance
  • Exceptionally comfortable lining: no seams on the working surfaces
  • Last longer than precision work gloves, yet offers better comfort and dexterity than heavy-duty gloves
Ideal for precision mechanical work, handling oily mechanical parts, wood working, packaging and manufacturing small electrical components.
Certificeringer: EN 388:2016 (3111X), Cat. II