General purpose gloves, Titan 328
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34328020 34328028 34328029
MAPA34328020EA 344 DKK
MAPA34328020 MAPA34328028 MAPA34328029
General purpose gloves, Titan 328
Handsker
These natural latex gloves with textile support provide flexibility and grip for common handling tasks.
  • Exceptionally comfortable fabric lining, and seamless knit
  • Good ventilation
  • Excellent grip, due to embossed texture
Ideal for household waste collection, warehousing and receipt of goods, handling bricks/blocks, plastering, public park maintenance and for handling materials when working on the roads.
Certificeringer: EN 388:2016 (2142X), EN 407 (X2XXXX); Cat. II