VWR® SAS Super DUO 360 Air Samplers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
710-0867EA 57370 DKK
710-0867 710-0866
VWR® SAS Super DUO 360 Air Samplers
Prøvetagere Luftprøvetager
When environmental contamination is very low, and the number of samples to be performed is high, operational duration can be an important factor in the choice of SAS instrument. Two sampling heads that operate simultaneously to sample 1000 litres of air, in less than 3 minutes, is the solution.
  • Single body with two aspirating heads
  • All manuals are available on a USB key
  • Instruction manual in six different languages
The simultaneous sampling on two heads also allows you to obtain statistically representative results when using the same type of media in both heads. It is also possible to use two different media for the same sampling cycle (TSA or PCA for Total Bacterial Count and SDA for yeasts and moulds). Sampling with the two heads can halve sample process time, which is a real advantage for busy users.
Certificeringer: ISO 14698 and USP 1116 Pharmacopoeia.
Bestillingsoplysninger: For det komplette system, bestil den sampler du har brug for, en batterioplader og det passende hoved.