VWR®, SAS Super ISO luftprøveudtagere
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
710-2087EA 47880 DKK
710-2087 710-2088 710-2089 710-2090
VWR®, SAS Super ISO luftprøveudtagere
Prøvetagere Luftprøvetager
SAS Super ISO 100 and SAS Super ISO 180 are differentiated by their airflow; this choice depends on the application, and allows continuous or sequential sampling from a few minutes to several hours. The data can be stored on the SAS instrument itself or on any remote PC.
  • Design avoids turbulence in unidirectional airflow and re-aspiration of tested air
  • Portable and easily positioned, they operate with long life, rechargeable batteries
  • All manuals are available on a USB key in six languages
  • Micro USB for data download and instrument configuration
  • New Data Management - CFR 21 Part 11 compliance
Derived from the SAS instrument used on the International Space Station, the SAS Super ISO has been specifically designed for pharmaceutical and hospital sectors.
The SAS SUPER ISO USB 100 and SAS SUPER ISO USB 180 air samplers are compliant with 21 CFR Part 11. They feature an electronic audit trail as well as a variety of security and monitoring functions for individual operators.
Certificeringer: ISO 14698 and USP 1116 Pharmacopoeia.
Bestillingsoplysninger: For det komplette system, bestil den sampler du har brug for, en batterioplader og det passende hoved.