Chemical resistant gloves, Butoflex 650 / Butoflex 651
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34650139 34650130 34650131 34650137 34651130
MAPA34650139EA 952 DKK
MAPA34650139 MAPA34650130 MAPA34650131 MAPA34650137 MAPA34651130
Chemical resistant gloves, Butoflex 650 / Butoflex 651
Handsker
These butyl gloves are ideal for the handling of very corrosive acids, ketones, esters and amine derivatives. Also suitable for metal treatment and degreasing.
  • Exclusive embossed exterior finish, meaning slippery objects can be handled without risk
  • Unrivalled flexibility and dexterity
  • Exceptional comfort thanks to textile lining (model 650 only)
  • In compliance with the requirement of the norm EN 16350, standard for protective gloves (model 651 only)
Available with and without a knitted textile interior.
Certificeringer: Model 650: EN 388:2016 (1121X), EN ISO 374-1 Type A (ABCILMNOS); Cat. III
Model 651: EN 388:2016 (0010X), EN ISO 374-1 Type A (ABCILMNOS); Cat. III.