QuEChERS dispersive SPE, Supra-d™
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9306910 N9306912 N9306921 N9306923 N9306927 N9306933 N9306900 N9306901 N9306902 N9306909
549-0719EA 3230 DKK
549-0719 549-0721 549-0725 549-0727 549-0731 549-0732 549-0714 549-0715 549-0716 549-0718
QuEChERS dispersive SPE, Supra-d™
Kromatografikolonner
QuEChERS (Quick, Easy, Cheap, Effective, Rugged, Safe) dispersive SPE, is a leading sample preparation approach in pesticide residue analysis. It eliminates complex liquid extraction methods and extends the range of recovered pesticides. The QuEChERS procedure is fast and easy, improving laboratory productivity, resulting in fewer errors.
  • High recoveries are achieved for more accurate analyses
  • Up to four times faster than traditional methods
  • Low solvent usage and waste for maximum cost savings
These Supra-d™ dispersive SPE extraction and clean-up kits have been customised for specific sample preparation needs. They are designed for both steps of the QuEChERS method and each kit includes pre-prepared products for simplicity and error-free extractions. A certificate of quality is included in each kit.