Industrial gloves, Alto 258
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34258008 34258006 34258007 34258329 34258000
112-2998EA 287 DKK
112-2998 112-2994 112-2997 112-2999 112-2993
Industrial gloves, Alto 258
Handsker
These liquidproof gloves made of natural latex provide a good resistance to many diluted acids.
  • Cotton flock-lining over an internal layer of white natural rubber
  • Curved fingers and contoured palm
  • Non-slip textured finish in palm and fingers area for improved grip
  • External surface lightly coated with silicone
Typical applications include: Work with composite materials (resins), common handling tasks, assembly of small, oil and grease free parts and minor maintenance tasks.
Thickness: 0,60 mm
Colour: Yellow
EN 388 (2120), EN 374; Cat. III; AQL 0,65 Food contact approved
Certificeringer: EN 388 (1110X), EN ISO 374-1 Type B (KPS), EN ISO 374-5; Cat. III