Latex gloves, Solo 998
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34998009 34998006 34998007 34998008
112-0778EA 346 DKK
112-0778 112-0775 112-0776 112-0777
Latex gloves, Solo 998
Handsker
These multipurpose, ambidextrous gloves with beaded cuffs are made of natural latex.
  • Non sterile, chlorinated, textured fingertips
  • Smooth with pebbled fingertips
  • Also suitable for common tasks in hospitals and clinics
Ideal for manufacturing of medicines, pharmaceutical preparation, research, analysis and handling of precision parts, as well as for the assembly of small, oil- and grease-free parts in the automotive industry.
Length: 300 mm
Thickness: 0,10 mm
Colour: Natural
EN 374 (Cat. III), AQL 1,5
Certificeringer: EN ISO 374-1 Type C, EN ISO 374-5; Cat. III; AQL 1,5