Ultrasonic baths, BactoSonic®
Leverandør: BANDELIN-ELECTRONIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3290 3291
BAND3290EA 36290 DKK
BAND3290 BAND3291
Ultrasonic baths, BactoSonic®
Ultralydsbad
Bactosonic gently removes the biofilms from implant surfaces.
  • Works with very low ultrasound intensity
  • Biofilm is removed without killing bacteria
  • Compare to standard methods up to 10000 times more bacteria can be detected
Leveringsinformation: Complete set includes ultrasonic bath, wire frame for foil test, TICKOPUR TR-3 concentrate (250 ml), implant boxes, and box trays.
Bestil nu
Specifikationer
Model BactoSonic® 14.2
Environmentally Preferable
Volumen 9,5 L
Frekvens 40 kHz
Tank W×D×H 325×300×150 mm
W×D×H 355×325×305 mm