Evening primrose oil
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3691.1 3691.2
ROTH3691.1EA 0 DKK
ROTH3691.1 ROTH3691.2
Evening primrose oil
Evening primrose oil
MDL Number: MFCD00131042
CAS-nummer: 90028-66-3
EINECS: 289-859-2