4-(Hydroxymethyl)pyrocatechol ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D2620-1G
D2620-5G
TCIAD2620-1GEA
747
DKK
TCIAD2620-1G
TCIAD2620-5G
4-(Hydroxymethyl)pyrocatechol ≥96.0% (by GC)
4-(Hydroxymethyl)pyrocatechol
Formel:
C₇H₈O₃ Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00210409 CAS-nummer: 3897-89-0 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 115.0 to 125.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...