4-Methylbenzyl isothiocyanate 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L11135.09 L11135.04
L11135.09EA 1210 DKK
L11135.09 L11135.04
4-Methylbenzyl isothiocyanate 96%
4-Methylbenzyl isothiocyanate

Purity: 96% CAS No.: 3694-46-0 Molecular Formula: C9H9NS Formula Weight: 163.24 Refractive Index: 1.5890 Odour: Pungent Boiling Point: 97°/1mm Beilstein Reference: 2082478 UN No.: 2922 MDL No.: MFCD00041118

Formel: C₉H₉NS
Kogepunkt: 97 °C (1 mmHg)
Massefylde: 1,080
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041118
CAS-nummer: 3694-46-0
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR