Methyl-4-methyl-3-nitrobenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 4-methyl-3-nitrobenzoate

M2146-25G M2146-5G
TCIAM2146-25GEA 559 DKK
TCIAM2146-25G TCIAM2146-5G
Methyl-4-methyl-3-nitrobenzoate ≥98.0% (by GC)
Methyl-4-methyl-3-nitrobenzoate
Formel: C₉H₉NO₄
MDL Number: MFCD00130054
CAS-nummer: 7356-11-8
EINECS: 230-886-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21a08fbae5b1ef08033c43f36def2faa01df2ad&vid=b21a08fbae5b1ef08033c43f36def2faa01df2ad&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR