Methyl-3-methyl-4-nitrobenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 3-methyl-4-nitrobenzoate

M2145-25G M2145-5G
TCIAM2145-25GEA 1010 DKK
TCIAM2145-25G TCIAM2145-5G
Methyl-3-methyl-4-nitrobenzoate ≥98.0% (by GC)
Methyl-3-methyl-4-nitrobenzoate
Formel: C₉H₉NO₄
MDL Number: MFCD00085640
CAS-nummer: 24078-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859903624bc55cf8b46e0e58a5e19725a228a2af&vid=859903624bc55cf8b46e0e58a5e19725a228a2af&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR