4-(Dimethylamino)phenyl isothiocyanate 97%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
L09011.09 L09011.04
L09011.09EA 3660 DKK
L09011.09 L09011.04
4-(Dimethylamino)phenyl isothiocyanate 97%
4-(Dimethylamino)phenyl isothiocyanate
Purity: 97% CAS No.: 2131-64-8 Molecular Formula: C9H10N2S Formula Weight: 178.25 Storage Temperature: 2°C to 8°C Odour: Irritating Boiling Point: 152°/6mm Beilstein Reference: 388270 UN No.: 2928 MDL No.: MFCD00041093
Formel: C₉H₁₀N₂S
Molekylvægt: 178.25 g/mol
Kogepunkt: 152 °C (6 mmHg)
Smeltepunkt: 64…70 °C
MDL Number: MFCD00041093
CAS-nummer: 2131-64-8
EINECS: 218-360-4
ADR: 6.1 II