3-(4-Morpholino)propyl isothiocyanate 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L12819.03 L12819.06
L12819.03EA 402 DKK
L12819.03 L12819.06
3-(4-Morpholino)propyl isothiocyanate 96%
3-(4-Morpholino)propyl isothiocyanate

Purity: 96% CAS No.: 32813-50-6 Molecular Formula: C8H14N2OS Formula Weight: 186.28 Storage Temperature: 2°C to 8°C Refractive Index: 1.5305 Boiling Point: 182-183°/13mm Beilstein Reference: 879217 UN No.: 2922 MDL No.: MFCD00041222

Formel: C₈H₁₄N₂OS
Kogepunkt: 182…183 °C (13 mmHg)
MDL Number: MFCD00041222
CAS-nummer: 32813-50-6
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR