4-Phenyl-1-butene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P0161-25ML
P0161-5ML
TCIAP0161-25MLEA
969
DKK
TCIAP0161-25ML
TCIAP0161-5ML
4-Phenyl-1-butene ≥98.0% (by GC)
4-Phenyl-1-butene
Formel:
C₁₀H₁₂ Kogepunkt: 181°C Flammepunkt: 53°C |
MDL Number:
MFCD00026111 CAS-nummer: 768-56-9 EINECS: 405-980-7 UN nummer: 3295 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...