3,5-Dimethylcyclohexanol (mixture of isomers) ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,5-Dimethylcyclohexanol

D0702-25ML D0702-500ML
TCIAD0702-25MLEA 468 DKK
TCIAD0702-25ML TCIAD0702-500ML
3,5-Dimethylcyclohexanol (mixture of isomers) ≥98.0% (by GC)
3,5-Dimethylcyclohexanol (mixture of isomers)
Formel: C₈H₁₆O
Molekylvægt: 128.2120 g/mol
MDL Number: MFCD00001447
CAS-nummer: 5441-52-1
EINECS: 226-630-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a961c49b4cdbb0095e34c4f02b4e5f60f9791633&vid=a961c49b4cdbb0095e34c4f02b4e5f60f9791633&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 % (total of isomer)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR