Tetrabutylammonium fluoride hydrate ≥98.0% (by titrimetric analysis) for catalyst of silylation, for cleavage of silyl ether

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Tetrabutylammonium fluoride xhydrate

T1037-100G T1037-25G
TCIAT1037-100GEA 2610 DKK
TCIAT1037-100G TCIAT1037-25G
Tetrabutylammonium fluoride hydrate ≥98.0% (by titrimetric analysis) for catalyst of silylation, for cleavage of silyl ether
Tetrabutylammonium fluoride hydrate
Formel: C₁₆H₃₆FN·H₂O
Molekylvægt: 279.4774 g/mol
MDL Number: MFCD00011747
CAS-nummer: 22206-57-1
EINECS: 207-057-2
UN nummer: 1759
ADR: 8 III
Merck Index: 14,09187
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d1a68e48d5d7af9e206fbdbfef15608ec42b6d0&vid=e01b3155302ebba9a8122dff2d77cb15e85d8e6e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 20.0 %
TBHF-HF (neutralization titration) max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR