4-Bromo-4'-hydroxybiphenyl ≥99.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4'-Bromo-[1,1'-biphenyl]-4-ol
H0690-25G H0690-5G
TCIAH0690-25GEA 1430 DKK
TCIAH0690-25G TCIAH0690-5G
4-Bromo-4'-hydroxybiphenyl ≥99.0% (by GC)
4-Bromo-4'-hydroxybiphenyl
Formel: C₁₂H₉BrO
Molekylvægt: 249.11 g/mol
MDL Number: MFCD00059076
CAS-nummer: 29558-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85688c2ded1f11ffe492635485db7fea09393e21&vid=85688c2ded1f11ffe492635485db7fea09393e21&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 167.0 to 170.0 deg-C
Solubility in Methanol almost transparency