(S)-(+)-Glycidyl butyrate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(S)-Oxiran-2-ylmethyl butyrate
G0283-25G
G0283-5G
TCIAG0283-25GEA
1240
DKK
TCIAG0283-25G
TCIAG0283-5G
(S)-(+)-Glycidyl butyrate ≥98.0% (by GC)
(S)-(+)-Glycidyl butyrate
Formel:
C₇H₁₂O₃ Molekylvægt: 144.1684 g/mol Flammepunkt: 84°C |
MDL Number:
MFCD00273369 CAS-nummer: 65031-96-1 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +29.0 to +33.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...