2-Aminoimidazole hemisulphate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)
A1665-1G A1665-5G
TCIAA1665-1GEA 132 DKK
TCIAA1665-1G TCIAA1665-5G
2-Aminoimidazole hemisulphate ≥98.0% (by HPLC, titration analysis)
2-Aminoimidazole hemisulphate
Formel: C₆H₁₂N₆O₄S
Molekylvægt: 264.26 g/mol
Smeltepunkt: 270°C
MDL Number: MFCD00013162
CAS-nummer: 1450-93-7
EINECS: 215-918-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3efff3c3db5254217afadd79b484f7e4389fa6dc&vid=3efff3c3db5254217afadd79b484f7e4389fa6dc&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Yellow to Orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %