HPLC columns, Hypersil GOLD™ SAX
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26305-013001 26305-014001 26303-052130 26305-154630 26305-104630 26302-152130 26302-102130 26302-052130 26305-254630 26303-154630
555-6380EA 4300 DKK
555-6380 555-6381 555-6530 555-6401 555-6396 555-6518 555-6516 554-4329 554-4331 554-4330
HPLC columns, Hypersil GOLD™ SAX
Kromatografikolonner
These quaternary amine strong anion-exchange columns are highly stable for aqueous and low pH mobile phases.
  • Analyse smaller organic molecules, including nucleotides and organic acids
  • Outstanding peak shape and sensitivity
Based on high purity silica bonded with an ion-exchange ligand, Hypersil GOLD™ SAX HPLC columns provide outstanding peak shape and sensitivity.
Bestillingsoplysninger: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2,1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4,6 mm int.Ø require the 4/4,6 mm UNIGUARD holder (10046-744).