Capillary columns, TraceGOLD™ TG-WaxMS B
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26086-1420 26086-1430 26086-2980 26086-2230
555-8938EA 7610 DKK
555-8938 555-8937 555-8930 555-8936
Capillary columns, TraceGOLD™ TG-WaxMS B
Kromatografikolonner
Base deactivation allows analysis of basic analytes without derivatization or column priming.
  • Polar phase, base deactivated polyethylene glycol phase
  • Reduced absorption and improved responsiveness for basic compounds
  • Not suitable for use with water or alcohols