Brix sugar standards for refractometers, Certipur®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5.00420.0001 5.00430.0001 5.00400.0001 5.00405.0001 5.00410.0001 5.00440.0001 5.00460.0001 5.00450.0001
5.00420.0001EA 2080 DKK
5.00420.0001 5.00430.0001 5.00400.0001 5.00405.0001 5.00410.0001 5.00440.0001 5.00460.0001 5.00450.0001
Brix sugar standards for refractometers, Certipur®, Supelco®
Standarder Brydningsindex Standarder
Refractive index standards traceable to SRM from NIST and PTB.
  • Each ampoule contains about 8 ml solution
  • Minimum shelf life of Brix standard 0° is 36 months
  • All other standards minimum shelf life is 24 month
  • Tested for homogeneity and stability
  • Storage at room temperature