Beakers, low form, stainless steel
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
075.100 075.250 075.500 075.005 075.003 075.002 075.001
213-1801EA 734 DKK
213-1801 213-1802 213-1803 RSGA075.005 RSGA075.003 RSGA075.002 RSGA075.001
Beakers, low form, stainless steel
Bægre
Stainless steel 18/10, with spout, straight edge.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.