4,4'-Isopropylidendicyclohexanol ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B0928-25G B0928-500G
TCIAB0928-25GEA 132 DKK
TCIAB0928-25G TCIAB0928-500G
4,4'-Isopropylidendicyclohexanol ≥93.0% (by GC)
4,4'-Isopropylidendicyclohexanol
Formel: C₁₅H₂₈O₂
MDL Number: MFCD00019334
CAS-nummer: 80-04-6
EINECS: 201-244-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4a4810cdd77e36da64ab816057c39dd154e1600&vid=b4a4810cdd77e36da64ab816057c39dd154e1600&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR