N-Benzylanthranilic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B2769-25G B2769-5G
TCIAB2769-25GEA 665 DKK
TCIAB2769-25G TCIAB2769-5G
N-Benzylanthranilic acid ≥98.0% (by titrimetric analysis)
N-Benzylanthranilic acid
Formel: C₁₄H₁₃NO₂
MDL Number: MFCD00020247
CAS-nummer: 6622-55-5
EINECS: 229-572-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c141eab533a37de4c071f1e5d99d304dfbfa6720&vid=c141eab533a37de4c071f1e5d99d304dfbfa6720&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 169.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR