2-Amino-6-methoxypyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A2228-1G
A2228-5G
TCIAA2228-1GEA
132
DKK
TCIAA2228-1G
TCIAA2228-5G
2-Amino-6-methoxypyridine ≥98.0% (by GC, titration analysis)
2-Amino-6-methoxypyridine
Formel:
C₆H₈N₂O Molekylvægt: 124.14 g/mol Kogepunkt: 115°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD04972542 CAS-nummer: 17920-35-3 |
|
Specifikationer Testresultater
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...