8-Nonenyl acetate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A1384-25ML
A1384-5ML
TCIAA1384-25MLEA
1660
DKK
TCIAA1384-25ML
TCIAA1384-5ML
8-Nonenyl acetate ≥98.0% (by GC)
8-Nonenyl acetate
Formel:
C₁₁H₂₀O₂ Kogepunkt: 155°C |
MDL Number:
MFCD00671492 CAS-nummer: 13038-22-7 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...