Tetraethylphosphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T1747-25G T1747-5G
TCIAT1747-25GEA 3000 DKK
TCIAT1747-25G TCIAT1747-5G
Tetraethylphosphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)
Tetraethylphosphonium tetrafluoroborate
Formel: C₈H₂₀BF₄P
MDL Number: MFCD01631314
CAS-nummer: 665-49-6
UN nummer: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=458b17b91499708eafe93cd6809fab25cea5802e&vid=458b17b91499708eafe93cd6809fab25cea5802e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR