Tetrakis(trimethylsilyl)silane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: TMSS

T1464-1G T1464-5G
TCIAT1464-1GEA 222 DKK
TCIAT1464-1G TCIAT1464-5G
Tetrakis(trimethylsilyl)silane ≥98.0% (by GC)
Tetrakis(trimethylsilyl)silane
Formel: C₁₂H₃₆Si₅
Molekylvægt: 320.842 g/mol
MDL Number: MFCD00054859
CAS-nummer: 4098-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859fc86f48ea09890d0bef8dbfedb2402a2657c5&vid=859fc86f48ea09890d0bef8dbfedb2402a2657c5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR