Isobornyl acetate ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

I0306-25ML I0306-500ML
TCIAI0306-25MLEA 132 DKK
TCIAI0306-25ML TCIAI0306-500ML
Isobornyl acetate ≥90.0% (by GC)
Isobornyl acetate
Formel: C₁₂H₂₀O₂
Molekylvægt: 196.29 g/mol
Kogepunkt: 106°C
Smeltepunkt: 7°C
Flammepunkt: 100°C
MDL Number: MFCD00135943
CAS-nummer: 125-12-2
EINECS: 204-727-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a196d75a86e8f3a28181c130650d5559be91b4e&vid=2a196d75a86e8f3a28181c130650d5559be91b4e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %
Specific rotation [a]20/D -2.0 to +2.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR