Diethyl aminomalonate hydrochloride

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A1016-25G A1016-500G A1016-100G
TCIAA1016-25GEA 140 DKK
TCIAA1016-25G TCIAA1016-500G TCIAA1016-100G
Diethyl aminomalonate hydrochloride
Diethyl aminomalonate hydrochloride
Formel: C₇H₁₄ClNO₄
MDL Number: MFCD00012510
CAS-nummer: 13433-00-6
EINECS: 236-556-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebb3e9d7e42de546bf3e61c722fa35eecf3b2af&vid=bebb3e9d7e42de546bf3e61c722fa35eecf3b2af&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR