(S)-(-)-3-Acetamidopyrrolidine ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
L12927.03
L12927.06
L12927.03EA
753
DKK
L12927.03
L12927.06
(S)-(-)-3-Acetamidopyrrolidine ≥98%
(S)-(-)-3-Acetamidopyrrolidine
Purity: 98%
CAS No.: 114636-31-6
Molecular Formula: C6H12N2O
Formula Weight: 128.17
Melting Point: 52-57°
Boiling Point: 70-72°/40mm
UN No.: 3259
MDL No.: MFCD00082748
Formel:
C₆H₁₂N₂O Kogepunkt: 70…72 °C (40 mmHg) Smeltepunkt: 52…57 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00082748 CAS-nummer: 114636-31-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...